Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Standard Pump Head Mounting Hardware, Mounts Two L/S 36 Pump Head, Stainless Steel; 2/Set
Catalog No. NC1417257
Encompass
Two end bell assemblies are required for a complete head assembly.
Catalog No. NC1417257 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0703602
Two end bell assemblies are required for a complete head assembly.