Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Standard Pump Head for Precision Tubing L/S 17, Polycarbonate Housing, SS Rotor
Catalog No. NC9618246
Encompass
This Standard pump head accepts precision tubing¿ ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 13, L/S 14, L/S 16, L/S 17, L/S 18.
Catalog No. NC9618246 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0701721
This Standard pump head accepts precision tubing¿ ensures optimal performance from your Masterflex pump. Select a pump head for one of the following precision tubing sizes: L/S 13, L/S 14, L/S 16, L/S 17, L/S 18.