Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P High-Performance Precision Pump Tubing, C-Flex, I/P 89; 10 ft
Catalog No. NC1341625
Encompass
Masterflex C-Flex High- Performance Precision pump tubing offers enhances biocompatibility with longer pumping life. This tubing is nonpyrogenic, noncytotoxic, and nonhemolytic ¿ protecting the purity of your fluid. Heat sealable and weldable, C-Flex tubing is ideal for biopharmaceutical processing and storage and botanical production.
Catalog No. NC1341625 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0642489/DEL
Masterflex C-Flex High- Performance Precision pump tubing offers enhances biocompatibility with longer pumping life. This tubing is nonpyrogenic, noncytotoxic, and nonhemolytic ¿ protecting the purity of your fluid. Heat sealable and weldable, C-Flex tubing is ideal for biopharmaceutical processing and storage and botanical production.