Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Bulk-Packed High-Performance Precision Pump Tubing, Platinum-Cured Silicone, L/S 15; 20 Bags of 25 ft/Bg
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9640415/DEL
Encompass
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent Our bulk tubing comes in convenient cases containing up to 20 sealed bags of tubing (depending on tubing size). Bulk tubing eliminates individual cardboard boxes, reduces unpacking time, and saves you money on shipping. Tubing comes in 25-ft bags.
Catalog No. NC9794164
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.