Learn More
AVANTOR MASTERFLEX (VWR LLC) Pump Tubing, Puri-Flex, B/T 91, 10 ft.
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9641991/DEL
Encompass
Masterflex Pump Tubing, Puri-Flex, B/T 91, 10 ft., Long life tubing for any Masterflex pump used in your pharma and bioprocess applicationsBiocompatible.Heat sealable and weldable.Long pump life when compared to silicone or other TPE tubings.Low spallation when compared to silicone or some other TPE tubings.Very low protein binding.
Catalog No. NC1420992