Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Tubing Loading Key for Standard Pump Heads
Catalog No. NC0192472
Encompass
L/S and I/P Standard pump heads include tubing loading keys. A key is required to ensure proper tubing alignment and tension. Order a replacement key based on your pump head series, L/S or I/P.
Catalog No. NC0192472 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0701390
L/S and I/P Standard pump heads include tubing loading keys. A key is required to ensure proper tubing alignment and tension. Order a replacement key based on your pump head series, L/S or I/P.