Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Sanitary Fitting Precision Pump Tubing Assembly, PharMed BPT, L/S 14; 10 ft
Catalog No. NC1662161
Encompass
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige Requires sanitary gaskets for both connections to ensure quality seal
This item is not returnable. View return policy
Catalog No. NC1662161 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9610314
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige Requires sanitary gaskets for both connections to ensure quality seal