Learn More
AVANTOR MASTERFLEX (VWR LLC) Chem-Durance Bio Pump Tubing, L/S 35, 50 ft.
Catalog No. NC1120521
Encompass
Masterflex L/S Chem-Durance Bio Pump Tubing, L/S 35, 50 ft, Applications: Laboratory, pharmaceutical processing, and biotech applications for pump lines.Characteristics: Excellent chemical resistance. Excellent life and durability under pressure with low spallation. Plasticizer-free inner liner with elastomer jacket. Opaque.
Catalog No. NC1120521 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0644235/DEL
Masterflex L/S Chem-Durance Bio Pump Tubing, L/S 35, 50 ft, Applications: Laboratory, pharmaceutical processing, and biotech applications for pump lines.Characteristics: Excellent chemical resistance. Excellent life and durability under pressure with low spallation. Plasticizer-free inner liner with elastomer jacket. Opaque.