Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Precision Pump Tubing, Tygon Fuel & Lubricant F-4040-A, L/S 25; 50 ft
Catalog No. NC9921942
Encompass
Applications: Fuels and industrial lubricants¿gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants Characteristics: Resists most hydrocarbons, embrittlement, and swelling; ozone- and UV-resistant, low extractability, thermoplastic, PVC-based material with plasticizer, firm (stiff) material, translucent yellow
Catalog No. NC9921942 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0640125
Applications: Fuels and industrial lubricants¿gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants Characteristics: Resists most hydrocarbons, embrittlement, and swelling; ozone- and UV-resistant, low extractability, thermoplastic, PVC-based material with plasticizer, firm (stiff) material, translucent yellow