Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Precision Pump Tubing, Peroxide-Cured Silicone, I/P 73; 150 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9640273
Encompass
Masterflex peroxide-cured silicone High-Performance Precision pump tubing features greater physical compression capability with a longer pumping life compared to platinum-cured silicone tubing. Translucent tubing is made with no additives or plasticizers and makes for easy flow monitoring. Ideal for general-purpose, biotech, and food and beverage applications, this tubing complies with USP Class VI, FDA, and 3-A standards.
Catalog No. NC0231687