Learn More
AVANTOR MASTERFLEX (VWR LLC) USB to DB9 (M) RS-232 Serial Adapter Cable
Catalog No. NC0762643
Encompass
Masterflex USB to DB9 (M) RS-232 Serial Adapter Cable, Use this cable to connect the Masterflex Computer Compatible drives 07550- and 07551- series to a PC with USB port. Will also connect other peripheral devices equipped with RS-232 serial interface including balances, printers, scanners, and PDA's. Includes 2-ft cable and driver software. Supports data transfer speeds up to 12 Mbps; compatible with Windows 2000 / XP / Vista / 7.
Catalog No. NC0762643 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX2205058
Masterflex USB to DB9 (M) RS-232 Serial Adapter Cable, Use this cable to connect the Masterflex Computer Compatible drives 07550- and 07551- series to a PC with USB port. Will also connect other peripheral devices equipped with RS-232 serial interface including balances, printers, scanners, and PDA's. Includes 2-ft cable and driver software. Supports data transfer speeds up to 12 Mbps; compatible with Windows 2000 / XP / Vista / 7.