Learn More
AVANTOR MASTERFLEX (VWR LLC) Tygon E-LFL Pump Tubing, L/S 24, 25 ft.
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0644024/DEL
Encompass
Masterflex L/S Tygon E-LFL Pump Tubing, L/S 24, 25 ftHigh-performance pump tubing for challenging applicationsNontoxic and nonhemolytic with excellent biocompatibilityLongest life of all Tygon formulationsPump at continuous pressures up to 40 psiLow extractables and low gas permeabilityMeets USP Class VI criteria and is FDA compliant. Applications: General laboratory use; provides longer life with peristaltic tubing pumps.Characteristics: Long life, won¿t age or oxidize, fits tightly to glass and metal, bends to small radius curves, nontoxic, and transparent.
Catalog No. NC0482433