Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Easy-Load II Pump Head for Precision Tubing, PPS Housing, CRS Rotor
Catalog No. NC9710829
Encompass
These pump heads accept the six L/S precision tubing sizes. For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately under "Accessories". When two Easy-Load II pump heads are stacked rollers are offset by 45¿--combine the outlets from two stacked heads for a 95% reduction in flow pulsation.
Catalog No. NC9710829 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7720050
These pump heads accept the six L/S precision tubing sizes. For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately under "Accessories". When two Easy-Load II pump heads are stacked rollers are offset by 45¿--combine the outlets from two stacked heads for a 95% reduction in flow pulsation.