Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P High-Performance Precision Pump Tubing, Platinum-Cured Silicone, I/P 88; 10 ft
Catalog No. NC0635187
Encompass
From a global market leader in range of formations, this tubing is distinguished by a smooth inner surface designed to reduce the risk of particle entrapment and contamination. Offering extensive chemical compatibility, genuine Masterflex tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment. Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility.
This item is not returnable. View return policy
Catalog No. NC0635187 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9651088/DEL
From a global market leader in range of formations, this tubing is distinguished by a smooth inner surface designed to reduce the risk of particle entrapment and contamination. Offering extensive chemical compatibility, genuine Masterflex tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment. Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility.