Learn More
Buehler MASTERPREP 0.05MICRON 64OZ
Supplier:  Buehler 406377064
Encompass
This item has a minimum qty of 3 per supplier requirements.
MASTERPREP Polishing Suspension, Particle size 0.05m, Steels tool steels, Ni-based alloys, Co-based alloys, Qty 64 oz.
Catalog No. NC9774560