Learn More
StatLab MasterPress Tissue Tampers
SDP
Catalog No. 21033473
Change view
Click to view available options
Width (Metric):
1.5 cm
1.7 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
21-033-473 1.7 cm
21-033-472 1.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-033-473 Supplier StatLab Supplier No. TCZ01201P
Add to Cart
Add to Cart
An excellent tool for ensuring embedded tissue is flat in the cassette mold
  • An essential tool in histology for ensuring that tissue specimens are properly oriented, evenly distributed, and tightly compacted within embedding media
  • For flattening tissue sections over the cooling molten paraffin into base molds
  • 3 Pk.
Specifications
Product Type Tissue Tampers
Quantity 3 Pk.
Length (Metric) 2 cm
Width (Metric) 1.7 cm