Learn More
StatLab MasterPress Tissue Tampers with Pink Handles
SDP
Catalog No. 21402413
Change view
Click to view available options
Includes:
Large Rectangle Tamper
Medium Rectangle Tamper
Round Tamper
Small Square Tamper
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
21-402-413 Large Rectangle Tamper
21-402-410 Round Tamper
21-402-411 Small Square Tamper
21-402-412 Medium Rectangle Tamper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-402-413 Supplier StatLab Supplier No. TCZ1106PKP
Add to Cart
Add to Cart
MasterPressTissue Tampers for Tissue Embedding
  • MasterPress Tissue Tampers help to ensure proper tissue orientation, even disribution, and tight compaction with embedding media
  • These tampers aid in the orientation and flatness for paraffin embedding
  • 2/Pack
Specifications
Product Type Tissue Tampers
Color Pink
Includes Large Rectangle Tamper
Quantity 2 per pack