Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 0.05% Lithium Carbonate
SDP

Catalog No. 22023759
Change view
Click to view available options
Quantity:
1 pt.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-759 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-759 Supplier StatLab Supplier No. KC2622
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab MasterTech 0.05% Lithium Carbonate - a bluing reagent in the staining process

  • Apply this after hematoxylin staining to remove excess hematoxylin and to provide a sharp contrast between the cell nuclei and the surrounding tissue
  • This differentiation improves the clarity and quality of microscopic images, allowing for precise analysis of tissue structure and morphology
  • A component for the MasterTech Luxol Fast Blue-Cresyl Echt Violet Stain Kit
  • A MasterTech Special Stain Kit Component

Specifications

Chemical Name or Material Lithium Carbonate
Physical Form Liquid or Solid
Quantity 1 pt.
Molecular Formula Li2CO3
Linear Formula Li2CO3
UN Number NONH for all modes of transport
Synonym dilithium carbonate
Solubility Information Fully Miscible
SMILES [Li+].[Li+].[O-]C(=O)[O-]
IUPAC Name dilithium carbonate
Concentration or Composition (by Analyte or Components) 0.05%
Manufacturing Origin USA
Product Line MasterTech
Vapor Pressure 23 hPa at 20°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.