Learn More
StatLab MasterTech 0.1% Gold Chloride
SDP
Catalog No. 21033266
Change view
Click to view available options
Product Type:
Gold Chloride
Catalog No. Product Type
21-033-266 Gold Chloride
1 options
Catalog No. 21-033-266 Supplier StatLab Supplier No. AHG0226
Add to Cart
Add to Cart
Used to stain the cell bodies
  • Used as a stain to visualize nerve fibers and neurons where it selectively binds to and highlights the delicate structures of neural tissue by reducing gold ions to visible metallic gold particles at the binding sites
  • A component used for the following MasterTech Stain Kits: Fontana Masson Stain Kit and Chandlers Precision Reticulum Stain Kit
  • A MasterTech Special Stain Kit Component
Specifications
Product Type Gold Chloride