Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MasterTech 0.1% Silver Nitrate Solution, StatLab
SDP

Catalog No. 22035370
Change view
Click to view available options
Quantity:
473 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-035-370 473 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-035-370 Supplier SLMP Llc Dba Statlab Medical Supplier No. AHS0843
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

Used as a staining agent for detecting specific tissue components through argentaffin or argyrophilic reactions.

  • A key reagent in silver staining techniques which rely on the reduction of silver ions to metallic silver to produce a black high-contrast image of tissue components
  • A StatLab MasterTech Special Stain Kit Component

Specifications

Quantity 473 mL
Product Type Silver Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.