Learn More
StatLab MasterTech 0.2% Fast Green Stain

Description
- Used to enhance the visibility of certain structures in histological specimens
- This stain is particularly useful for differentiating various types of tissues and for identifying specific cellular components that may not be visible under standard staining techniques
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| Chemical Name or Material | Fast Green Stain |
| CAS | 64-17-5, 67-63-0 |
| Color | Green |
| Quantity | 16 oz. |
| Percent Purity | 0.2% |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.