Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MasterTech 0.3% Gold Chloride Solution, StatLab
SDP

Catalog No. 22024783
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-024-783 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-024-783 Supplier SLMP Llc Dba Statlab Medical Supplier No. KC0921
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as a toning agent to stabilize silver stains by converting unstable metallic silver into stable metallic gold.

  • It increases contrast, improves image clarity, and prevents staining from fading
  • A StatLab MasterTech Special Stain Kit Component

Specifications

Physical Form Solution
Color Golden/Green
Quantity 100 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.