Learn More
StatLab MasterTech 0.3% Gold Chloride
SDP
Catalog No. 21010303
Change view
Click to view available options
Quantity:
1 Pint
Catalog No. Quantity
21-010-303 1 Pint
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-303 Supplier StatLab Supplier No. KC0922
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Used for staining fungi and Pneumocystis jirovecii.
  • A component for the MasterTech GMS Stain Kit
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 1 Pint
Product Type Gold Chloride