Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 0.5% Toluidine Blue O Alcohol
SDP

Catalog No. 21010283
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-010-283 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-283 Supplier StatLab Supplier No. CUTBA.5PT
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

It used for staining various histological tissues and cells.

  • Stain is particularly useful for highlighting metachromatic elements, which are substances that exhibit different colors when viewed under a microscope
  • The dye binds to negatively charged molecules in the cell, such as nucleic acids and acidic proteins, allowing for the visualization of nuclei, mast cell granules, and cartilage matrix
  • A MasterTech Special Stain Kit Component

Specifications

Chemical Name or Material Toluidine Blue Stain
CAS 64-17-5
Color Blue
pH Unknown
Quantity 1 Pint
Molecular Weight (g/mol) Unknown
Percent Purity 2.50%
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.