Learn More
StatLab MasterTech 1% Acid Alcohol

Description
- Differentiates bacteria from background tissue by removing excess Advance Carbol Fuchsin Stain used in the MasterTech Acid Fast Bacteria Stain Kit and the Fites Stain Kit procedures
- Used to remove excess stain and define nuclei following hematoxylin staining in the HandE and PAP methods
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| Quantity | 1 L |
| Product Type | 1% Focus Acid Alcohol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.