Learn More
StatLab MasterTech 1% Alcian Blue
SDP
Catalog No. 22021470
Change view
Click to view available options
Quantity:
1 Pint
Catalog No. Quantity
22-021-470 1 Pint
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-470 Supplier StatLab Supplier No. KC4172
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Used to stain acidic epithelial mucins, such as sialomucin and sulfomucin, as well as stromal (mesenchymal) mucins.
  • Stain is particularly useful in diagnosing conditions like Barrett's esophagus, mesothelioma, atherosclerosis, and adenocarcinomas
  • The dye binds to the negatively charged sulfur and/or carboxyl groups of mucins, depending on the pH used, allowing for the visualization of acidic mucins in tissue samples
  • A component of the MasterTech Russell Movat Special Stain Kit
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 1 Pint
Product Type Alcian Blue