Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 1% Lithium Carbonate
SDP

Catalog No. 22023768
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-768 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-768 Supplier StatLab Supplier No. AHL0250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab 1% Lithium Carbonate, A MasterTech Special Stain Kit Component

  • 1% Lithium Carbonate is used as a bluing agent for histology and cytology staining
  • It is particularly effective in the LFB/CEV stain and is part of the bluing reagents used for histology staining
  • The solution is mixed with distilled water and stored at room temperature
  • It is applied after hematoxylin staining and clearing/differentiation to achieve the desired blue color in the tissue samples
  • A MasterTech Special Stain Kit Component

Specifications

Concentration 1%
Quantity 1 L
Product Type Bluing Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.