Learn More
StatLab MasterTech 1% Potassium Permanganate

Description
- Used as a bleaching agent to decolorize melanin pigment in tissue sections allowing for better visualization of underlying tissue details and is also sometimes used as a differentiating agent to distinguish between different types of amyloid deposits
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| Concentration | 1% |
| For Use With (Equipment) | Staining Equipment |
| Grade | Chemical Solution |
| Product Type | Imaging Reagent |
| Quantity | 1 pt. |
| Solution Type | Stain Kit |
| Form | Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.