Learn More
StatLab MasterTech 1% Safranin O Solution

Description
- Solution provides a brilliant red to orange-red coloration to these components, offering excellent contrast with other stains
- It is particularly useful in bone and joint pathology, as well as in studying connective tissue disorders and inflammatory processes
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| Quantity | 16 oz. |
| Product Type | Safranin O |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.