Learn More
MasterTech 1% Sodium Bisulfite, StatLab

Description
- It acts as a mild reducer to remove excess permanganate stains (used in oxidization steps) from tissues
- In procedures involving bleaching with chlorine or similar compounds, sodium bisulfite serves as an antichlor to remove residual oxidizers
- A StatLab MasterTech Special Stain Kit Component
Specifications
Specifications
| Physical Form | Solution |
| CAS | 7631-90-5 |
| Color | Colorless |
| Quantity | 100 mL |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.