Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 1% Toluidine Blue O Stain
SDP

Catalog No. 22021423
Change view
Click to view available options
Quantity:
1 pt.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-423 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-423 Supplier StatLab Supplier No. STTBOPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab MasterTech 1% Toluidine Blue O Stain - used for staining various acidic structures in cells

  • It is particularly useful for highlighting metachromatic elements, substances that exhibit different colors when viewed under a microscope
  • The stain binds to negatively charged molecules in the cell, such as nucleic acids and acidic proteins, allowing for the visualization of nuclei, mast cell granules, and cartilage matrix
  • This staining technique is valuable in medical research and diagnostics, providing insights into the structure and composition of tissues and cells.
  • A MasterTech Special Stain Kit Component

Specifications

Chemical Name or Material TOLUIDINE BLUE O STAIN
Concentration 1%
Grade Histological
Color Blue
Quantity 1 pt.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.