Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 10% Alcoholic Hematoxylin
SDP

Catalog No. 22033331
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-033-331 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-033-331 Supplier StatLab Supplier No. KC4112
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to make the working Verhoeffs Elastic Stain within the MasterTech Russell-Movat Pentachrome Stain Kit

  • This stain is essential for making the working Verhoeffs Elastic Stain, which is used to assess the elasticity of tissues
  • The stain is part of a series of stains that help in the diagnosis and analysis of various pathological conditions
  • A component of the MasterTech Russell-Movat Pentachrome Stain Kit
  • A MasterTech Special Stain Kit Component

Specifications

For Use With (Equipment) Staining Equipment
Grade Chemical Solution
Product Type Hematoxylin
Quantity 16 oz.
Solution Type Stain
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.