Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 10% Ferric Chloride
SDP

Catalog No. 21010253
Change view
Click to view available options
Color:
Brown
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-010-253 Brown
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-253 Supplier StatLab Supplier No. AHF0126
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Used in the Verhoeff elastin stain, which is a histological technique for staining elastic fibers.

  • This stain is particularly useful in the study of connective tissues, such as ligaments and tendons, where elastin fibers are present
  • The Verhoeff stain allows pathologists to visualize the architecture of these tissues and differentiate between different types of connective tissues
  • A component for the following MasterTech Stain Kits - Russell-Movat Pentachrome Stain Kit and Verhoeffs Elastic Stain Kit
  • A MasterTech Special Stain Kit Component

Specifications

Physical Form Solution
CAS 10025-77-1
Color Brown
Quantity 100 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.