Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 10% Neutral Buffered Formalin
SDP

Catalog No. 22021362
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-362 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-362 Supplier StatLab Supplier No. FXBFOLT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab MasterTech 10% Neutral Buffered Formalin (NBF) - quality NBF for proper fixation

  • StatLab MasterTech 10% NBF is the most trusted fixative for all Pathology applications
  • MasterTech 10% Buffered Formalin is ready to use and buffered to pH 7.0 for optimal specimen fixation, transport and processing
  • An effective fixative for H and E, most immunohistochemistry (IHC) markers and special stains
  • Manufactured to strict quality assurance standards to eliminate machinery-clogging precipitate

Specifications

Concentration 10%
For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Formalin
Quantity 1 L
Solution Type Formalin
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.