Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MasterTech 10% Silver Nitrate Solution, StatLab
SDP

Catalog No. 22024779
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-024-779 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-024-779 Supplier SLMP Llc Dba Statlab Medical Supplier No. AHS1043
Add to Cart
Add to Cart

Used as a Reagent in Staining.

  • Used as a reagent in staining procedures to highlight specific tissue components, such as reticular fibers, nerves, and pathogens, by forming visible metallic silver deposits
  • Highlights connective tissue fibers to study structural architecture
  • A StatLab MasterTech Special Stain Kit Component

Specifications

Physical Form Solution
CAS 7761-88-8
Color Clear Colorless
Quantity 500 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.