Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MasterTech 10% Zinc Formalin Solution, StatLab
SDP

Catalog No. 22035373
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-035-373 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-035-373 Supplier SLMP Llc Dba Statlab Medical Supplier No. FXZFO100
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

A superior, non-coagulating histology fixative.

  • Used to enhance nuclear and cytoplasmic detail acting as a rapid safer alternative to mercury-based fixatives
  • Fixative improves antigen preservation for immunohistochemistry IHC especially for lymphoid markers and increases tissue rigidity for better sectioning

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Fixative
Quantity 100 mL
Solution Type Formalin
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.