Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 12% Acetic Acid
SDP

Catalog No. 22021415
Change view
Click to view available options
Quantity:
1 pt.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-415 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-415 Supplier StatLab Supplier No. KC0342
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab MasterTech 12% Acetic Acid - a component for the MasterTech Colloidal Iron Stain Kit

  • Used as a component in fixative solutions where it helps preserve nuclei by precipitating DNA counteracting shrinkage caused by other fixative ingredients like ethanol and ensuring good tissue penetration while minimizing the loss of nucleic acids
  • A MasterTech Special Stain Kit Component

Specifications

Concentration 12%
For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Tissue Preservative
Quantity 1 pt.
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.