Learn More
StatLab MasterTech 2.5% Gum Mastic Solution
SDP
Catalog No. 22033319
Change view
Click to view available options
Form:
Liquid
Catalog No. Form
22-033-319 Liquid
1 options
This item is not returnable. View return policy
Catalog No. 22-033-319 Supplier StatLab Supplier No. AHG0343
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for the demonstration of spirochetes, such as those associated with Lyme disease, and other nonfilamentous bacteria
  • Solution is prepared by dissolving gum mastic in absolute alcohol and is then filtered through Whatman 4 filter paper and is stable for six months when stored in a refrigerator
  • The solution is used in conjunction with other reagents to create a reducing solution that facilitates the staining process
  • A component for the following MasterTech Stain Kits: Steiner Stain Kit, Dieterles Stain Kit, and the Modified Genta Triple Stain Kit
  • A MasterTech Special Stain Kit Component
Specifications
Form Liquid
Product Type Gum Mastic Solution