Learn More
MasterTech 2% Sodium Thiosulfate, StatLab

Description
- It acts as a reducing agent to stabilize color, remove non-specific staining, and improve contrast in nerve fibers or bacteria visualization
- A StatLab MasterTech Special Stain Kit Component
Specifications
Specifications
| Physical Form | Solution |
| CAS | 7772-98-7 |
| Color | Colorless |
| Quantity | 500 mL |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.