Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 20% Formalin
SDP

Catalog No. 22021377
Change view
Click to view available options
Quantity:
1 pt.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-377 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-377 Supplier StatLab Supplier No. KC3942
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab MasterTech 20% Formalin - used as a tissue fixative to preserve the tissue structure before staining

  • Used as a tissue fixative to preserve the tissue structure before staining ensuring the reticular fibers within the tissue are properly preserved and visible under a microscope when stained with the kits specific reagents
  • A MasterTech Special Stain Kit Component
  • A component for the MasterTech Chandlers Precision Reticulum Stain Kit

Specifications

Concentration 20%
For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Fixative
Quantity 1 pt.
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.