Learn More
StatLab MasterTech 3% Acetic Acid
SDP
Catalog No. 21033264
Change view
Click to view available options
Product Type:
Acid Alcohol (3% HCl)
Catalog No. Product Type
21-033-264 Acid Alcohol (3% HCl)
1 options
Catalog No. 21-033-264 Supplier StatLab Supplier No. AHA0443
Add to Cart
Add to Cart
Used to adjust the pH of tissue sections
  • Used to adjust the pH of tissue sections, particularly when performing Alcian blue staining, as the optimal staining result depends on the acidic pH created by the acetic acid solution
  • The use of a 3% acetic acid solution helps to adjust the pH of tissue sections before or after staining to ensure the binding of Alcian Blue to the desired molecules
  • A component for the following MasterTech Stain Kits: Alcian Blue pH1.0, Alcian Blue pH2.5, and the Russell-Movat Pentachrome Stain Kit
  • A MasterTech Special Stain Kit Component
Specifications
Product Type Acid Alcohol (3% HCl)