Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 3% Potassium Ferrocyanide
SDP

Catalog No. 21010257
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-010-257 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-257 Supplier StatLab Supplier No. AHP0450
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

A solution that can be used in the Colloidal Iron staining procedure.

  • This compound is essential for enhancing the visibility of iron-containing components in tissue samples, which is crucial for accurate histological analysis
  • The solution is yellow, odorless, and is used in various laboratory settings for its ability to clearly visualize cellular structures and other components within the tissue samples
  • A component of the MasterTech Colloidal Iron Stain Kit
  • A MasterTech Special Stain Kit Component

Specifications

Quantity 1 L
Product Type Potassium Ferrocyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.