Learn More
StatLab MasterTech 3% Potassium Ferrocyanide
SDP
Catalog No. 22031872
Change view
Click to view available options
Quantity:
1 gal.
Catalog No. Quantity
22-031-872 1 gal.
1 options
Catalog No. 22-031-872 Supplier StatLab Supplier No. AHP0457
Add to Cart
Add to Cart
For detecting iron deposits in tissue samples.
  • Used as the key reagent in the Perl's Prussian Blue stain to detect and visualize the presence of ferric iron in biological tissues
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 1 gal.
Product Type Potassium Ferrocyanide