Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 3% Potassium Metabisulfite
SDP

Catalog No. 21010307
Change view
Click to view available options
Quantity:
1 Pint
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-010-307 1 Pint
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-307 Supplier StatLab Supplier No. KC3922
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

A primary Component of the Reticulum Stain Kit in Histology.

  • This stain is a crucial component in the reticulum staining process, which is a method used to visualize the reticular endoplasmic reticulum in tissues
  • The reticulum stain is essential for identifying various cellular structures and is used in conjunction with other stains to provide a comprehensive view of the tissue's morphology and cellular composition
  • A component for the MasterTech Chandlers Precision Reticulum Stain Kit
  • A MasterTech Special Stain Kit Component

Specifications

Physical Form Solution
CAS 16731-55-8
Color Colorless
Quantity 1 Pint
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.