Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MasterTech 3% Potassium Metabisulfite, StatLab
SDP

Catalog No. 22024790
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-024-790 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-024-790 Supplier SLMP Llc Dba Statlab Medical Supplier No. KC3921
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used as a reducing agent in specialized staining techniques.

  • It stabilizes samples by inhibiting oxidative damage and acts as a bleaching agent to remove background, ensuring accurate results
  • It acts as a 'rinse' or reducing agent after Schiff reagent application, helping to remove excess reagent and prevent background coloration, particularly in detecting glycogen or mucins
  • A StatLab MasterTech Special Stain Kit Component

Specifications

Physical Form Solution
CAS 16731-55-8
Color Colorless
Quantity 100 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.