Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 3% Sodium Hydroxide
SDP

Catalog No. 21033271
Change view
Click to view available options
Grade:
Reagent
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
21-033-271 Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-271 Supplier StatLab Supplier No. AHS0643
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A highly alkaline solution that helps in the microscopic examination of biological samples

  • Used as a tissue digestive agent, specifically to help dissolve and separate tissue components, like connective tissue fibers, to allow for better visualization of the cellular structures within a tissue sample during the preparation process for microscopic examination
  • Solution is used to prepare stains that allow for the visualization of cell and tissue components
  • A component for the following MasterTech Stain Kits: HelicoStat Rapid Helicobacter Stain Kit and Chandlers Precision Reticulum Stain Kit
  • A MasterTech Special Stain Kit Component

Specifications

Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
Quantity 16 oz.
Percent Purity 3%
Grade Reagent
Packaging Plastic Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.