Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 3% Sodium Hydroxide
SDP

Catalog No. 21010261
Change view
Click to view available options
Physical Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Physical Form
21-010-261 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-261 Supplier StatLab Supplier No. AHS0650
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

A highly alkaline solution that helps in the visualization of cell and tissue components.

  • It is employed to oxidize tissue sections, allowing for the visualization of melanin, which is a brown pigment found in hair, skin, and other tissues
  • The staining process involves a series of steps, including the use of potassium permanganate to oxidize the tissue, followed by decolorization and counterstaining with haematoxylin to reveal the nuclei
  • A component for the following MasterTech Stain Kits Chandlers Precision Reticulum Stain Kit
  • A MasterTech Special Stain Kit Component

Specifications

Physical Form Solution
CAS 1310-73-2
Color Clear Colorless
Quantity 1 L
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.