Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 4% Paraformaldehyde Phosphate Buffer
SDP

Catalog No. 21010263
Change view
Click to view available options
Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-010-263 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-263 Supplier StatLab Supplier No. BUP0157
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

A crucial fixative in histology, used to preserve cellular structures and maintain tissue integrity for detailed analysis.

  • Solution is particularly effective in immunohistochemistry and fluorescent protein-labeled samples
  • The solution is prepared by dissolving paraformaldehyde in PBS, which helps prevent polymerization and maintains a neutral pH to avoid the formation of formic acid
  • A MasterTech Special Stain Kit Component

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Quantity 1 gal.
Solution Type Paraformaldehyde
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.