Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 5% Chromic Acid
SDP

Catalog No. 22021409
Change view
Click to view available options
Quantity:
1 pt.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-409 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-409 Supplier StatLab Supplier No. KC0912
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

StatLab MasterTech 5% Chromic Acid - used as a fixative to preserve tissue structures and morphology

  • Used as a fixative particularly for staining fungal elements in a procedure like the Grocott-Gomori methenamine silver (GMS) stain
  • A MasterTech Special Stain Kit Component

Specifications

Concentration 5%
For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Fixative
Quantity 1 pt.
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.