Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech 5% Sodium Thiosulfate
SDP

Catalog No. 21033270
Change view
Click to view available options
Grade:
Chemical Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
21-033-270 Chemical Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-270 Supplier StatLab Supplier No. AHS0326
Add to Cart
Add to Cart

Used as a stop-bath to remove excess silver ions from tissue sections after silver staining techniques

  • Solution can also be used to remove iodine when treating mercury-treated slides
  • It can also be used to detect argentophilic and argentaffin fibers in histology samples
  • A component for the following MasterTech Stain Kits: Bodian 2-Hour Microwave Stain Kit, Fontana-Masson Stain Kit, Chandlers Precision Reticulum Stain Kit, Russell-Movat Pentachrome Stain Kit, Verhoeff's Elastic Stain Kit, and Von Kossa Stain Kit
  • A MasterTech Special Stain Kit Component

Specifications

For Use With (Equipment) Staining Equipment
Grade Chemical Solution
Product Type Sodium Thiosulfate
Quantity 100 mL
Solution Type Stop Bath
For Use With (Application) Manual Special Staining
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.